Skip to Content
Merck
All Photos(1)

Key Documents

771759

Sigma-Aldrich

2-Methylpyridine borane complex solution

0.5 M in dichloromethane

Synonym(s):

2-Picoline borane complex solution, 2-Picoline borane complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H7N · BH3
CAS Number:
Molecular Weight:
106.96
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

0.5 M in dichloromethane

mp

44-46 °C (lit.)

density

1.297 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[BH3-][n+]1ccccc1C

InChI

1S/C6H10BN/c1-6-4-2-3-5-8(6)7/h2-5H,1,7H3

InChI key

JWXUMNOOAYRPKH-UHFFFAOYSA-N

Application

  • Environmentally benign reducing agent

Reactant for:
  • Reductive amination
  • N-Benzyl-protection of amino acid derivatives by reductive alkylation
  • Reductive alkoxyamination

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service