Skip to Content
Merck
All Photos(1)

Key Documents

769355

Sigma-Aldrich

Zinc sulfide

sublimed, granular, 1-4 mm

Synonym(s):

Zinc sulphide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnS
CAS Number:
Molecular Weight:
97.46
EC Number:
MDL number:
UNSPSC Code:
12161600

form

granular
sublimed

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

particle size

1-4 mm, coeff var ≥80%
1-4 mm

density

4.1 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.001%
Cr: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.002%
Hg: ≤0.1%
Pb: ≤0.1%
V: ≤0.005%

SMILES string

S=[Zn]

InChI

1S/S.Zn

InChI key

WGPCGCOKHWGKJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Uses:
  • Preparation of flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cells
  • Prepare a composite CdS-ZnS/Zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible light
  • Prepare light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dots
  • Catalyst for photocatalytic degradation of organic pollutants
  • Preparation of color tunable light-emitting diodes (LEDs)
  • Prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysis
  • Catalyst for synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael addition
  • Prepare CdSe/ZnS q uantum dots for chemiluminescent and chemiluminescence resonance energy transfer (CRET) detection of DNA, metal ions, and aptamer-substrate complexes
  • Preparation of ZnS nanocrystals for ultrasensitive protein detection in terms of multiphonon resonance Raman scattering
Zinc sulfide finds applications in anti-reflection coating. The use of anti-reflection coating using ZnS has been known to increase the effeiciency of solar cells as well.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

34.1 °F - Not applicable

Flash Point(C)

1.185 °C - Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-thin GaAs single-junction solar cells integrated with a reflective back scattering layer
Yang, W., et al.
Journal of Applied Physics, 115, 203105-203105 (2014)
Investigation on Antireflection Coatings for Silicon Solar Cells
Xue, C., et al.
Applied Mechanics and Materials, 521, 33-36 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service