Skip to Content
Merck
All Photos(3)

Key Documents

764485

Sigma-Aldrich

2-[2-(tert-Butyldimethylsilyloxy)ethoxy]ethanamine

95%

Synonym(s):

2-[2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]ethoxy]ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H25NO2Si
CAS Number:
Molecular Weight:
219.40
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.440

density

0.887 g/mL at 25 °C

functional group

amine

storage temp.

2-8°C

SMILES string

NCCOCCO[Si](C)(C(C)(C)C)C

InChI

1S/C10H25NO2Si/c1-10(2,3)14(4,5)13-9-8-12-7-6-11/h6-9,11H2,1-5H3

InChI key

BFDRSKJOABFLHC-UHFFFAOYSA-N

Application

Novel tert--butyl dimethylsilyl (TBDMS) protected PEG amine linker.

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

214.0 °F

Flash Point(C)

101.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service