Skip to Content
Merck
All Photos(1)

Key Documents

761915

Sigma-Aldrich

Methyl 4-bromoquinoline-6-carboxylate

95%

Synonym(s):

4-Bromo-6-quinolinecarboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€51.87

€51.87

List price€74.10Save 30%

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€51.87

About This Item

Empirical Formula (Hill Notation):
C11H8BrNO2
CAS Number:
Molecular Weight:
266.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€51.87

List price€74.10Save 30%

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

mp

145-150 °C

functional group

bromo
ester

SMILES string

COC(=O)c1ccc2nccc(Br)c2c1

InChI

1S/C11H8BrNO2/c1-15-11(14)7-2-3-10-8(6-7)9(12)4-5-13-10/h2-6H,1H3

InChI key

ROTWWLOBDCESBP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service