Skip to Content
Merck
All Photos(1)

Key Documents

761451

Sigma-Aldrich

1,3,8,10(2H,9H)-Tetraone, 2,9-bis(2-phenylethyl)anthra[2,1,9-def:6,5,10-d′e′f′]diisoquinoline

98% (elemental analysis)

Synonym(s):

PEPTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H26N2O4
CAS Number:
Molecular Weight:
598.65
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98% (elemental analysis)

form

solid

mp

415-420 °C

semiconductor properties

N-type (mobility=1.4 cm2/V·s)

SMILES string

O=C1N(CCc2ccccc2)C(=O)c3ccc4c5ccc6C(=O)N(CCc7ccccc7)C(=O)c8ccc(c9ccc1c3c49)c5c68

InChI

1S/C40H26N2O4/c43-37-29-15-11-25-27-13-17-31-36-32(40(46)42(39(31)45)22-20-24-9-5-2-6-10-24)18-14-28(34(27)36)26-12-16-30(35(29)33(25)26)38(44)41(37)21-19-23-7-3-1-4-8-23/h1-18H,19-22H2

InChI key

IIKSFQIOFHBWSO-UHFFFAOYSA-N

General description

1,3,8,10(2H,9H)-Tetraone, 2,9-bis(2-phenylethyl)anthra[2,1,9-def:6,5,10-d′e′f′]diisoquinoline (PEPTC) is an n-type small molecular organic semiconducting material. They are most commonly used in organic thin film transistors (OTFT) and also in solution processed organic photovoltaic cells (OPV). They have a well-defined structure and do not show any molecular weight dependence, leading to high purity and ease of processability.[1]

Application

Used as an n-type semiconducting material in organic and printed electronics.[1]

Features and Benefits

Organized nanostructure and high carrier mobility.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of small molecule donors for solution-processed organic solar cells
Payne, A. J., & Welch, G. C.
Material Matters, 9(1), 23-32 (2014)

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service