Skip to Content
Merck
All Photos(1)

Key Documents

761427

Supelco

α,α,α-Trifluorotoluene solution

NMR reference standard, 0.05% in chloroform-d ("100%", 99.96 atom % D), NMR tube size 5 mm × 8 in.

Synonym(s):

Benzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CF3
CAS Number:
Molecular Weight:
146.11
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

concentration

0.05% in chloroform-d ("100%", 99.96 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

FC(F)(F)c1ccccc1

InChI

1S/C7H5F3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H

InChI key

GETTZEONDQJALK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

19F sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hygienic standardization of benzotrifluoride level in reservoir water].
V I Antonova et al.
Gigiena i sanitariia, (2)(2), 86-87 (1985-02-01)
Shota Tsukada et al.
Neuropharmacology, 48(4), 479-491 (2005-03-10)
Glutamate transporters rapidly take up synaptically released glutamate and maintain the glutamate concentration in the synaptic cleft at a low level. (2S, 3S)-3-[3-[4-(trifluoromethyl)benzoylamino]benzyloxy]aspartate (TFB-TBOA) is a novel glutamate transporter blocker that potently suppresses the activity of glial transporters. TFB-TBOA inhibited
Bhawani Singh Rathore et al.
Bioorganic & medicinal chemistry, 14(16), 5678-5682 (2006-05-03)
7-Chloro-5-trifluoromethyl/7-fluoro/7-trifluoromethyl-4H-1,4-benzothiazines have been synthesized by 2-amino-5-fluoro/5-trifluoromethyl/5-chloro-3-trifluoromethyl benzenethiols condensed with beta-diketone/beta-ketoesters in the presence of DMSO involving oxidative cyclization. Pharmacological activities have also been included.
Hidetoshi Yamada et al.
Carbohydrate research, 337(7), 581-585 (2002-03-23)
A glycosylation reaction induced by copper(II) trifluoromethanesulfonate is described. Using benzotrifluoride as the reaction solvent, five kinds of glycosyl donors, a glucosyl chloride, a fluoride, a trichloroacetimidate, a 1-O-acetyl compound, and a lactol were activated to give the corresponding glucosides.
Eun Jin Cho et al.
Science (New York, N.Y.), 328(5986), 1679-1681 (2010-06-26)
The trifluoromethyl group can dramatically influence the properties of organic molecules, thereby increasing their applicability as pharmaceuticals, agrochemicals, or building blocks for organic materials. Despite the importance of this substituent, no general method exists for its installment onto functionalized aromatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service