Skip to Content
Merck
All Photos(1)

Documents

761249

Sigma-Aldrich

2-(Penta-4-ynyl)-2-oxazoline

96%

Synonym(s):

2-(Penta-4-ynyl)-4,5-dihydrooxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO
CAS Number:
Molecular Weight:
137.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

96%

form

liquid

refractive index

n20/D 1.476

density

1.005 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C#CCCCC1=NCCO1

InChI

1S/C8H11NO/c1-2-3-4-5-8-9-6-7-10-8/h1H,3-7H2

InChI key

MNUUNDKDTPELEI-UHFFFAOYSA-N

Application

Biocompatible multifunctional monomer:
  • This monomer can be used for click/ligation chemistry, as well as polymerized with other oxazolines.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

196.0 °F

Flash Point(C)

91.11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service