Skip to Content
Merck
All Photos(1)

Documents

760013

Sigma-Aldrich

3-Hydroxybenzisoxazole

97%

Synonym(s):

1,2-Benzisoxazol-3-ol, 1,2-Benzisoxazolin-3-one, 3-Hydroxy-1,2-benzisoxazole, Benzo[d]isoxazol-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO2
CAS Number:
Molecular Weight:
135.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

140-145 °C

SMILES string

Oc1noc2ccccc12

InChI

1S/C7H5NO2/c9-7-5-3-1-2-4-6(5)10-8-7/h1-4H,(H,8,9)

InChI key

QLDQYRDCPNBPII-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henry Sershen et al.
Neurochemical research, 41(1-2), 398-408 (2016-02-10)
Deficits in N-methyl-D-aspartate receptor (NMDAR) function are increasingly linked to persistent negative symptoms and cognitive deficits in schizophrenia. Accordingly, clinical studies have been targeting the modulatory site of the NMDA receptor, based on the decreased function of NMDA receptor, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service