Skip to Content
Merck
All Photos(1)

Documents

759155

Sigma-Aldrich

1,3-Di(2-pyridyl)-1,3-propanedione

97%

Synonym(s):

1,3-Bis(2-pyridyl)-1,3-propanedione, 1,3-Di-2-pyridinyl-1,3-propanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O2
CAS Number:
Molecular Weight:
226.23
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

104-109 °C

SMILES string

O=C(CC(=O)c1ccccn1)c2ccccn2

InChI

1S/C13H10N2O2/c16-12(10-5-1-3-7-14-10)9-13(17)11-6-2-4-8-15-11/h1-8H,9H2

InChI key

DCGUVLMWGIPVDP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service