755419
1,6,7,8-Tetrahydrocyclopenta[g]indole-3-carboxaldehyde
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
210-215 °C
SMILES string
[H]C(C1=CNC2=C1C=CC3=C2CCC3)=O
InChI
1S/C12H11NO/c14-7-9-6-13-12-10-3-1-2-8(10)4-5-11(9)12/h4-7,13H,1-3H2
InChI key
XMOYOVNKXCLBQX-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service