Skip to Content
Merck
All Photos(1)

Key Documents

753114

Sigma-Aldrich

(Acetonitrile)[2-di-tert-butyl(2′,4′,6′-triisopropylbiphenyl)phosphine]gold(I) hexafluoroantimonate

Synonym(s):

tBuXPhos Au(MeCN)SbF6, tert-Butyl XPhos Au(MeCN)SbF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H48AuF6NPSb
CAS Number:
Molecular Weight:
898.41
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

252-258 °C

SMILES string

[Au+].CC#N.F[Sb-](F)(F)(F)(F)F.CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c2ccccc2P(C(C)(C)C)C(C)(C)C

InChI

1S/C29H45P.C2H3N.Au.6FH.Sb/c1-19(2)22-17-24(20(3)4)27(25(18-22)21(5)6)23-15-13-14-16-26(23)30(28(7,8)9)29(10,11)12;1-2-3;;;;;;;;/h13-21H,1-12H3;1H3;;6*1H;/q;;+1;;;;;;;+5/p-6

InChI key

PYTYQNZDJXLXIE-UHFFFAOYSA-H

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service