Skip to Content
Merck
All Photos(1)

Documents

741701

Sigma-Aldrich

Ethyl 2-methyl-2-(phenylthiocarbonylthio)propionate

95%

Synonym(s):

2-(Ethoxycarbonyl)-2-propyldithiobenzoate, ECPDB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O2S2
CAS Number:
Molecular Weight:
268.39
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

refractive index

n20/D 1.600

density

1.148 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)(C)SC(=S)c1ccccc1

InChI

1S/C13H16O2S2/c1-4-15-12(14)13(2,3)17-11(16)10-8-6-5-7-9-10/h5-9H,4H2,1-3H3

InChI key

MDFLMFRGXQRZII-UHFFFAOYSA-N

Application

RAFT agent for controlled radical polymerization; well-suited for polymerization of methacrylates, methacrylamides, and to a lesser extent styrenes, acrylates, and acrylamides. Chain Transfer Agent (CTA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service