Skip to Content
Merck
All Photos(1)

Key Documents

741418

Sigma-Aldrich

1,3-Dimethyl-2-phenyl-2,3-dihydro-1H-benzoimidazole

97%

Synonym(s):

DMBI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3
CAS Number:
Molecular Weight:
267.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

mp

106-111 °C

semiconductor properties

N-type (mobility>10−2 cm2/V·s) (from a 2 wt% N-DMBI doped PCBM transistor)

SMILES string

CN1C(C2=CC=C(N(C)C)C=C2)N(C)C3=C1C=CC=C3

InChI

1S/C17H21N3/c1-18(2)14-11-9-13(10-12-14)17-19(3)15-7-5-6-8-16(15)20(17)4/h5-12,17H,1-4H3

InChI key

AKIIMLCQTGCWQQ-UHFFFAOYSA-N

General description

This material is shown to behave as a n-type dopant for n-channel Organic Thin Film Transistors (OTFTs).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Wei et al.
Journal of the American Chemical Society, 132(26), 8852-8853 (2010-06-18)
We present here the development of a new solution-processable n-type dopant, N-DMBI. Our experimental results demonstrated that a well-known n-channel semiconductor, [6,6]-phenyl C(61) butyric acid methyl ester (PCBM), can be effectively doped with N-DMBI by solution processing; the film conductivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service