Skip to Content
Merck
All Photos(1)

Key Documents

740632

Sigma-Aldrich

N-Tosyl-5-methyl-3,4-hexadienylamine

95%

Synonym(s):

4-Methyl-N-(5-methyl-3,4-hexadien-1-yl)benzenesulfonamide, N-Toluenesulfonyl-5-methyl-3,4-hexadienylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO2S
CAS Number:
Molecular Weight:
265.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

50-55 °C

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)=C=CCCNS(=O)(=O)c1ccc(C)cc1

InChI

1S/C14H19NO2S/c1-12(2)6-4-5-11-15-18(16,17)14-9-7-13(3)8-10-14/h4,7-10,15H,5,11H2,1-3H3

InChI key

LSSMBRKXEAJPMI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service