Skip to Content
Merck
All Photos(1)

Key Documents

735965

Sigma-Aldrich

Lauric acid-11,11,12,12,12-d5

98 atom % D, 98% (CP)

Synonym(s):

Dodecanoic acid-11,11,12,12,12-d5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2(CH2)9COOH
Molecular Weight:
205.35
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Assay

98% (CP)

mp

44-46 °C (lit.)

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])CCCCCCCCCC(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i1D3,2D2

InChI key

POULHZVOKOAJMA-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service