Skip to Content
Merck
All Photos(2)

Key Documents

734683

Sigma-Aldrich

Zirconium(IV) oxide-yttria stabilized

pellets (for sputtering)

Synonym(s):

YSZ-8, YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO2 · Y2O3
CAS Number:
Molecular Weight:
349.03
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

pellets (for sputtering)

composition

(ZrO2)0.92(Y2O3)0.08

diam. × H

2-3 mm × 3-5 mm

mp

>2,600 °C (lit.)

SMILES string

O=[Zr]=O.O=[Y]O[Y]=O

InChI

1S/5O.2Y.Zr

InChI key

KSTOPEBVPBSOTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Yttria-stabilized zirconia (YSZ) is the most commonly used solid electrolyte for solid oxide fuel cells (SOFCs) and oxygen sensors. YSZ exhibits high oxygen-ionic conductivity while blocking electronic conduction. YSZ particles may be synthesized from yttrium nitrate or isopropoxide by sol gel method. YSZ nanoparticles may be prepared in supercritical water (SCW). YSZ nanoparticles of different sizes with varying % mol of yttria may be obtained by CO2 laser based continuous/pulsed laser vaporization method.

Application

Collagen-yttria-stabilized amorphous zirconia hybrid scaffolds may be developed. YSZ thin films may be used as thermal barrier coatings on combustors, stationary and rotating components of gas turbines or diesel engines.6
Electrolyte material for solid oxide fuel cells (SOFC).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kowalski, K.
Diffusion and Defect Data, Solid State Data, Part A, 286-286 (2009)
Synthesis and dispersion of yttria-stabilized zirconia (YSZ) nanoparticles in supercritical water
Gonzalo-Juan I, et al.
Materials Chemistry and Physics, 134(1), 451-458 (2012)
Oxygen reduction via grain boundary transport in thin film platinum electrodes on yttria stabilized zirconia
Huber TM, et al.
Solid State Ionics, 273, 8-12 (2015)
Elisabeth W Leib et al.
Journal of colloid and interface science, 448, 582-592 (2015-03-22)
Zirconia microparticles produced by sol-gel synthesis have great potential for photonic applications. To this end, identifying synthetic methods that yield reproducible control over size uniformity is important. Phase transformations during thermal cycling can disintegrate the particles. Therefore, understanding the parameters
Impedance spectroscopy of pellets made from yttria stabilized zirconia nanoparticles generated via CW and pulsed mode of laser vaporization method
Khare J, et al.
Ceramics International, 40(9) Part B, 14677-14685 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service