Skip to Content
Merck
All Photos(1)

Key Documents

733377

Sigma-Aldrich

4-Vinylbenzocyclobutene

97%

Synonym(s):

3-Ethenyl(bicyclo[4.2.0]-octa-1,3,5-triene), VBCB

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€202.00

€202.00


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
€202.00

About This Item

Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€202.00


Estimated to ship on26 March 2025


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.578

density

0.962 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccc2CCc2c1

InChI

1S/C10H10/c1-2-8-3-4-9-5-6-10(9)7-8/h2-4,7H,1,5-6H2

InChI key

DTGDMPJDZKDHEP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

169.0 °F

Flash Point(C)

76.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service