Skip to Content
Merck
All Photos(1)

Key Documents

728020

Sigma-Aldrich

N-Boc-trans-4-(p-tosyloxy)-L-proline methyl ester

≥98% (HPLC)

Synonym(s):

(2S,4R)-4-{[(4-Methylphenyl)sulfonyl]oxy]-1,2-pyrrolidinedicarboxylic acid 1-(1,1-dimethylethyl) 2-methyl ester, (4R)-4-TsO-N-Boc-Pro-OMe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO7S
CAS Number:
Molecular Weight:
399.46
Beilstein:
7231334
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

composition

carbon, 53.3-54.9%
nitrogen, 3.2-3.8%

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COC(=O)[C@@H]1C[C@H](CN1C(=O)OC(C)(C)C)OS(=O)(=O)c2ccc(C)cc2

InChI

1S/C18H25NO7S/c1-12-6-8-14(9-7-12)27(22,23)26-13-10-15(16(20)24-5)19(11-13)17(21)25-18(2,3)4/h6-9,13,15H,10-11H2,1-5H3/t13-,15+/m1/s1

InChI key

UKVKNGDXVREPDE-HIFRSBDPSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorthe Skovgaard et al.
PloS one, 6(2), e16678-e16678 (2011-02-25)
Protein turnover in collagen rich tissue is influenced by exercise, but can only with difficulty be studied in vivo due to use of invasive procedure. The present study was done to investigate the possibility of applying the PET-tracer, cis-[(18)F]fluoro-proline (cis-Fpro)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service