Skip to Content
Merck
All Photos(1)

Documents

722960

Sigma-Aldrich

Glyceryl tri(oleate-2,3,7,8-13C4)

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C12C45H104O6
Molecular Weight:
897.34
UNSPSC Code:
12142200

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

mass shift

M+12

storage temp.

−20°C

SMILES string

O=C(O[13CH2][13CH2]CCC[13CH2][13CH2]/C=C/CCCCCCCC)CC(C(O[13CH2][13CH2]CCC[13CH2][13CH2]/C=C/CCCCCCCC)=O)CC(O[13CH2][13CH2]CCC[13CH2][13CH2]/C=C/CCCCCCCC)=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service