Skip to Content
Merck
All Photos(2)

Key Documents

720445

Sigma-Aldrich

1-Benzyl-1H-pyrazole-4-boronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BN2O2
CAS Number:
Molecular Weight:
202.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

mp

76-81 °C

storage temp.

2-8°C

SMILES string

OB(O)c1cnn(Cc2ccccc2)c1

InChI

1S/C10H11BN2O2/c14-11(15)10-6-12-13(8-10)7-9-4-2-1-3-5-9/h1-6,8,14-15H,7H2

InChI key

JXNAIAOHYPDQQC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service