Skip to Content
Merck
All Photos(1)

Documents

719358

Sigma-Aldrich

Sulfur-33S

99 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
33S
Molecular Weight:
32.97
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:

isotopic purity

99 atom %

form

solid

mp

119 °C (lit.)

mass shift

M+1

SMILES string

[33S]

InChI

1S/S/i1+1

InChI key

NINIDFKCEFEMDL-OUBTZVSYSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service