Skip to Content
Merck
All Photos(1)

Key Documents

719013

Sigma-Aldrich

(Piperidinium-1-ylmethyl)trifluoroborate

internal salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13BF3N
CAS Number:
Molecular Weight:
166.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

134-149 °C

SMILES string

F[B-](F)(F)C[NH+]1CCCCC1

InChI

1S/C6H12BF3N/c8-7(9,10)6-11-4-2-1-3-5-11/h1-6H2/q-1/p+1

InChI key

PFMNVTAJBBIDDU-UHFFFAOYSA-O

Application

(Piperidinium-1-ylmethyl)trifluoroborate can be used as a reagent in the palladium-catalyzed Suzuki-cross coupling reaction with aryl and heteroaryl halides/mesylates to prepare methylpiperidinyl derivatives.
Organotrifluoroborate involved in Suzuki-Miyaura cross-coupling and cross coupling with mesylated phenol derivatives

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reinvestigation of aminomethyltrifluoroborates and their application in Suzuki- Miyaura cross-coupling reactions
Raushel J, et al.
The Journal of Organic Chemistry, 76(8), 2762-2769 (2011)
Cross-coupling of mesylated phenol derivatives with potassium ammonio-and amidomethyltrifluoroborates
Molander GA and Beaumard F
Organic Letters, 13(5), 1242-1245 (2011)

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service