Skip to Content
Merck
All Photos(2)

Key Documents

717312

Sigma-Aldrich

2,5-Diaminoterephthalic acid

greener alternative

≥95% (HPLC)

Synonym(s):

H2DATA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2O4
CAS Number:
Molecular Weight:
196.16
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95% (HPLC)

form

solid

mp

>300 °C

greener alternative category

SMILES string

Nc1cc(c(N)cc1C(O)=O)C(O)=O

InChI

1S/C8H8N2O4/c9-5-1-3(7(11)12)6(10)2-4(5)8(13)14/h1-2H,9-10H2,(H,11,12)(H,13,14)

InChI key

WIOZZYWDYUOMAY-UHFFFAOYSA-N

General description

2,5-Diaminoterephthalic acid is used as a linker for the formation of metal organic frameworks (MOFs).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander von Tesmar et al.
Cell chemical biology, 24(10), 1216-1227 (2017-09-12)
In vitro reconstitution and biochemical analysis of natural product biosynthetic pathways remains a challenging endeavor, especially if megaenzymes of the nonribosomal peptide synthetase (NRPS) type are involved. In theory, all biosynthetic steps may be deciphered using mass spectrometry (MS)-based analyses of
Selected Applications of Metal-Organic Frameworks in Sustainable Energy Technologies?.
Caskey SR and Matzger AJ.
Material Matters, 4(4), 111-111 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service