Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.521
density
1.206 g/mL at 25 °C
SMILES string
CCCCCCCCc1ccsc1Br
InChI
1S/C12H19BrS/c1-2-3-4-5-6-7-8-11-9-10-14-12(11)13/h9-10H,2-8H2,1H3
InChI key
ISONQKSIWXLJOQ-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.
Organic electronics promise renewable energy solutions surpassing silicon-based tech.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service