Skip to Content
Merck
All Photos(1)

Key Documents

706523

Sigma-Aldrich

Chlorotris(trimethylsilyl)silane solution

50 wt. % in toluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H27ClSi4
CAS Number:
Molecular Weight:
283.11
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

form

liquid

concentration

50 wt. % in toluene

density

0.877 g/mL at 25 °C

SMILES string

C[Si](C)(C)[Si](Cl)([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C9H27ClSi4/c1-11(2,3)14(10,12(4,5)6)13(7,8)9/h1-9H3

InChI key

HCOROOJWVHFMRJ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service