Skip to Content
Merck
All Photos(1)

Key Documents

701076

Sigma-Aldrich

2,2-Difluorobutyric acid

97%

Synonym(s):

2,2-Difluorobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6F2O2
CAS Number:
Molecular Weight:
124.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.365

density

1.215 g/mL at 25 °C

SMILES string

CCC(F)(F)C(O)=O

InChI

1S/C4H6F2O2/c1-2-4(5,6)3(7)8/h2H2,1H3,(H,7,8)

InChI key

SBSKRVZFWQPJID-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Zivkovic et al.
The FEBS journal, 287(4), 800-813 (2019-09-06)
Isoleucyl-tRNA synthetase (IleRS) is a paradigm for understanding how specificity against smaller hydrophobic substrates evolved in both the synthetic and editing reactions. IleRS misactivates nonproteinogenic norvaline (Nva) and proteinogenic valine (Val), with a 200-fold lower efficiency than the cognate isoleucine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service