Skip to Content
Merck
All Photos(1)

Documents

694703

Sigma-Aldrich

Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine

Synonym(s):

Bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]phosphinous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46ClO2P
CAS Number:
Molecular Weight:
505.11
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

116-120 °C

functional group

phosphine

SMILES string

COc1c(cc(cc1C(C)(C)C)P(Cl)c2cc(c(OC)c(c2)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C30H46ClO2P/c1-27(2,3)21-15-19(16-22(25(21)32-13)28(4,5)6)34(31)20-17-23(29(7,8)9)26(33-14)24(18-20)30(10,11)12/h15-18H,1-14H3

InChI key

VOGHMZHRQUWLRE-UHFFFAOYSA-N

Application

Reactant for:
  • Asymmetric hydrogenation with iridium catalysts
  • Condensations with optically active diols
  • Chemoselective reaction with deprotonated 2-hydoxy-3,3,N-trimethylbutanamide

Reactant for synthesis of:
  • Phosphine-containing amino acids and peptide-based catalyst ligands
  • Nonracemic phosphine-containing amino acids for palladium-catalyzed asymmetric allylic substitution reactions

Legal Information

Product of Kanata Chemical Technologies, Inc.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service