Skip to Content
Merck
All Photos(1)

Documents

689726

Sigma-Aldrich

(R)-3-(Aminomethyl)-1-Boc-pyrrolidine

≥95.0% (HPLC)

Synonym(s):

(R)-3-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, tert-Butyl (R)-3-(aminomethyl)-1-pyrrolidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
Beilstein:
10537871
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

liquid

optical activity

[α]/D -24.0±1.0°, c = 0.5 in chloroform

refractive index

n20/D 1.477-1.479

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC[C@H](CN)C1

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-5-4-8(6-11)7-12/h8H,4-7,11H2,1-3H3/t8-/m1/s1

InChI key

OGCCBDIYOAFOGK-MRVPVSSYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service