Skip to Content
Merck
All Photos(1)

Documents

687715

Sigma-Aldrich

p-Tolyl diiodomethyl-13C sulfone

99 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC7H8I2O2S
Molecular Weight:
423.01
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

95% (CP)

form

solid

mass shift

M+1

SMILES string

CC1=CC=C(S(=O)([13C](I)I)=O)C=C1

InChI

1S/C8H7I2O2S/c1-6-2-4-7(5-3-6)13(11,12)8(9)10/h2-5H,1H3/i8+1

InChI key

NIERGPOMYDWQNZ-VJJZLTLGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Gastro-intestinal system,Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service