Skip to Content
Merck
All Photos(1)

Key Documents

686492

Sigma-Aldrich

3-Mercapto-N-nonylpropionamide

95%

Synonym(s):

C9-1AT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NOS
CAS Number:
Molecular Weight:
231.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

41-49 °C

storage temp.

−20°C

SMILES string

CCCCCCCCCNC(=O)CCS

InChI

1S/C12H25NOS/c1-2-3-4-5-6-7-8-10-13-12(14)9-11-15/h15H,2-11H2,1H3,(H,13,14)

InChI key

CKRXZOZYQYDWIQ-UHFFFAOYSA-N

General description

3-Mercapto-N-nonylpropionamide is an amine containing alkanethiol that forms a self-assembled monolayer (SAM) on a variety of surfaces. The amide groups incorporated into the alkylthiolated backbone provide robust, stable and chemically inert monolayers.[1][2]

Application

3-Mercapto-N-nonylpropionamide can be coated on gold surfaces for the fabrication of molecular scale devices.[1][2][3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase separation within a binary self-assembled monolayer on Au 111 driven by an amide-containing alkanethiol
Smith RK, et al.
The Journal of Physical Chemistry B, 105(6), 1119-1122 (2001)
Control and placement of molecules via self-assembly
Lewis PA, et al.
Nanotechnology, 12(3), 231-231 (2001)
Structural manipulation of hydrogen-bonding networks in amide-containing alkanethiolate monolayers via electrochemical processing
Kim M, et al.
The Journal of Physical Chemistry C, 114(46), 19744-19751 (2010)
Journal of the American Chemical Society, 121, 5319-5327 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service