Skip to Content
Merck
All Photos(1)

Key Documents

684260

Sigma-Aldrich

Dodecyltrimethyl-d34 ammonium bromide

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D34NBr
Molecular Weight:
342.55
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

solid

mol wt

341.87 by atom % calculation

mass shift

M+34

SMILES string

[Br-].[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2;

InChI key

XJWSAJYUBXQQDR-DIHFYDAMSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service