Skip to Content
Merck
All Photos(1)

Documents

682543

Sigma-Aldrich

2-Bromo-5-fluoroanisole

99%

Synonym(s):

1-Bromo-4-fluoro-2-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BrFO
CAS Number:
Molecular Weight:
205.02
Beilstein:
3240536
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.542

bp

143-145 °C/760 mmHg

density

1.5983 g/mL at 25 °C

SMILES string

COc1cc(F)ccc1Br

InChI

1S/C7H6BrFO/c1-10-7-4-5(9)2-3-6(7)8/h2-4H,1H3

InChI key

KGYXKRGMSUHYCY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F

Flash Point(C)

90 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service