Skip to Content
Merck
All Photos(1)

Key Documents

681296

Sigma-Aldrich

N-Boc-piperidine-2-methanol

97%

Synonym(s):

2-Hydroxymethylpiperidine-1-carboxylic acid tert-butyl ester, N-Boc-2-(hydroxymethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO3
CAS Number:
Molecular Weight:
215.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

74-78 °C

SMILES string

CC(C)(C)OC(=O)N1CCCCC1CO

InChI

1S/C11H21NO3/c1-11(2,3)15-10(14)12-7-5-4-6-9(12)8-13/h9,13H,4-8H2,1-3H3

InChI key

PZTAGFCBNDBBFZ-UHFFFAOYSA-N

Application

Reactant for:
  • Julia coupling for synthesis of corydendramine A
  • Cyclization-functionalization of nitrogen-containing dienes

Reactant for synthesis of:
  • Heterocyclic compounds
  • Anthranilamide inhibitors of factor Xa
  • Human GnRH receptor antagonists
  • Sphingosine-1-phosphate receptor agonists

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service