Recommended Products
Assay
97%
refractive index
n20/D 1.5042
bp
75-80 °C/450 mmHg
density
1.6343 g/mL at 25 °C
SMILES string
ClCC(Br)=C
InChI
1S/C3H4BrCl/c1-3(4)2-5/h1-2H2
InChI key
LGQUWHWIEUSLLO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
99.0 °F - closed cup
Flash Point(C)
37.2 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service