All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H2BrN
CAS Number:
Molecular Weight:
131.96
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
96%
refractive index
n20/D 1.482
bp
48-54 °C/150 mbar
density
1.595 g/mL at 25 °C
SMILES string
BrC(=C)C#N
InChI key
CMPIGRYBIGUGTH-UHFFFAOYSA-N
Other Notes
May form precipitate upon standing which does not affect use
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.1 °F - closed cup
Flash Point(C)
41.7 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service