Skip to Content
Merck
All Photos(1)

Key Documents

669172

Sigma-Aldrich

(S)-(+)-3,3′-Bis(3,5-dimethylphenyl)-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-bi-2-naphthol

97%

Synonym(s):

(S)-3,3′-bis(3,5-dimethylphenyl)-5,6,7,8,5′,6′,7′,8′-octahydro-[1,1′]binaphthalenyl-2,2′-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H38O2
Molecular Weight:
502.69
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

218-222 °C

SMILES string

Cc1cc(C)cc(c1)-c2cc3CCCCc3c(c2O)-c4c(O)c(cc5CCCCc45)-c6cc(C)cc(C)c6

InChI

1S/C36H38O2/c1-21-13-22(2)16-27(15-21)31-19-25-9-5-7-11-29(25)33(35(31)37)34-30-12-8-6-10-26(30)20-32(36(34)38)28-17-23(3)14-24(4)18-28/h13-20,37-38H,5-12H2,1-4H3

InChI key

RSHMDLJRFPHHRE-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

TADDOLs demonstrate versatility in metal-catalyzed asymmetric reactions and as Brønsted acid organocatalysts in hetero-Diels–Alder reactions.

We present an article concerning BINOL and Derivatives.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service