Skip to Content
Merck
All Photos(1)

Key Documents

669164

Sigma-Aldrich

(N-Boc-2-pyrrolyl)dimethylsilanol

97%

Synonym(s):

N-Boc-2-(hydroxydimethylsilyl)pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3COC(O))(C4H3N)(Si(CH3)2(OH))
CAS Number:
Molecular Weight:
241.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

48-57 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)n1cccc1[Si](C)(C)O

InChI

1S/C11H19NO3Si/c1-11(2,3)15-10(13)12-8-6-7-9(12)16(4,5)14/h6-8,14H,1-5H3

InChI key

RLDVDBYGGISJNL-UHFFFAOYSA-N

Application

(N-Boc-2-pyrrolyl)dimethylsilanol can be used as a substrate:
  • To prepare cross-coupling products by reacting with various aryl iodides.[1][2]
  • In the synthesis of alkoxyhydrosiloxanes by reacting with alkoxyhydrosilanes.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed cross-coupling reactions of heterocyclic silanolates with substituted aryl iodides and bromides
Denmark SE and Baird JD
Organic Letters, 8(4), 793-795 (2006)
A catalyst-and additive-free synthesis of alkoxyhydrosiloxanes from silanols and alkoxyhydrosilanes
Satoh Y, et al.
Chemical Communications (Cambridge, England), 56(59), 8218-8221 (2020)
Organosilicon Chemistry
Denmark SE and Baird JD
Aldrich Chemfiles, 6(5), 1-14 (2006)
Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Articles

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service