Skip to Content
Merck
All Photos(2)

Key Documents

668842

Sigma-Aldrich

(2R,5R)-(+)-5-Benzyl-3-methyl-2-(5-methyl-2-furyl)-4-imidazolidinone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2O2
CAS Number:
Molecular Weight:
270.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5598

storage temp.

2-8°C

SMILES string

CN1[C@@H](N[C@H](Cc2ccccc2)C1=O)c3ccc(C)o3

InChI

1S/C16H18N2O2/c1-11-8-9-14(20-11)15-17-13(16(19)18(15)2)10-12-6-4-3-5-7-12/h3-9,13,15,17H,10H2,1-2H3/t13-,15-/m1/s1

InChI key

IQIMPHPFMHVWBZ-UKRRQHHQSA-N

Application

Highly selective organocatalyst for the Diels-Alder reaction with simple α,β−unsaturated ketones.

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burk, M. J. et al
Journal of the American Chemical Society, 118, 5142-5142 (1996)

Articles

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service