Skip to Content
Merck
All Photos(1)

Documents

664790

Sigma-Aldrich

3-(4-Methylphenyl)benzaldehyde

97%

Synonym(s):

4′-Methyl-3-formylbiphenyl, 4′-Methyl[1,1′-biphenyl]-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O
CAS Number:
Molecular Weight:
196.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.6242

density

1.0758 g/mL at 25 °C

SMILES string

Cc1ccc(cc1)-c2cccc(C=O)c2

InChI

1S/C14H12O/c1-11-5-7-13(8-6-11)14-4-2-3-12(9-14)10-15/h2-10H,1H3

InChI key

VEEXIHQTCQUWED-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service