Skip to Content
Merck
All Photos(1)

Key Documents

663751

Sigma-Aldrich

2-[4-(Trifluoromethoxy)phenyl]ethanol

98%

Synonym(s):

2-(4-Trifluoromethoxyphenyl)ethanol, 4-(Trifluoromethoxy)benzeneethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9F3O2
CAS Number:
Molecular Weight:
206.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

83-86 °C/7.0 mmHg

density

1.2774 g/mL at 25 °C

SMILES string

OCCc1ccc(OC(F)(F)F)cc1

InChI

1S/C9H9F3O2/c10-9(11,12)14-8-3-1-7(2-4-8)5-6-13/h1-4,13H,5-6H2

InChI key

RILZRCJGXSFXNE-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service