Skip to Content
Merck
All Photos(1)

Key Documents

662887

Sigma-Aldrich

3-Methyl-1-hexyne

technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€661.00

€661.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€661.00

About This Item

Linear Formula:
HC≡CCH(CH3)CH2CH2CH3
CAS Number:
Molecular Weight:
96.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€661.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

technical grade

refractive index

n20/D 1.402

density

0.710 g/mL at 25 °C

SMILES string

CCCC(C)C#C

InChI

1S/C7H12/c1-4-6-7(3)5-2/h2,7H,4,6H2,1,3H3

InChI key

OPZULQHRFNTFFZ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service