Skip to Content
Merck
All Photos(1)

Key Documents

655732

Sigma-Aldrich

4-Hydroxy-1-piperidineethanol

96%

Synonym(s):

1-(2-Hydroxyethyl)-4-hydroxypiperidine, 4-Hydroxy-1-piperidineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO2
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

33-40 °C

SMILES string

OCCN1CCC(O)CC1

InChI

1S/C7H15NO2/c9-6-5-8-3-1-7(10)2-4-8/h7,9-10H,1-6H2

InChI key

DOILPQLHDAWKCZ-UHFFFAOYSA-N

Application

Reactant for synthesis of estrogen receptor ligands and breast cancer cell antiproliferative agents

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service