Skip to Content
Merck
All Photos(1)

Documents

645397

Sigma-Aldrich

3-Chloro-4-(4′-fluorobenzyloxy)phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BClFO3
CAS Number:
Molecular Weight:
280.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

181-186 °C (lit.)

SMILES string

OB(O)c1ccc(OCc2ccc(F)cc2)c(Cl)c1

InChI

1S/C13H11BClFO3/c15-12-7-10(14(17)18)3-6-13(12)19-8-9-1-4-11(16)5-2-9/h1-7,17-18H,8H2

InChI key

GLLLLVYDIZNTKE-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service