Skip to Content
Merck
All Photos(1)

Documents

641901

Sigma-Aldrich

Yttrium(III) oxide, dispersion

nanoparticles, <100 nm particle size (BET), 5 wt. % in H2O, ≥99.9% trace metals basis

Synonym(s):

Yttria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y2O3
CAS Number:
Molecular Weight:
225.81
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.9% trace metals basis

form

nanoparticles

contains

1.0 wt. % Disperbyk®-190 as dispersant

concentration

5 wt. % in H2O

particle size

<100 nm (BET)
<50 nm (XRD)

pH

7-9

conductivity

10-100 μS/cm

SMILES string

O=[Y]O[Y]=O

InChI

1S/3O.2Y

InChI key

SIWVEOZUMHYXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DISPERBYK is a registered trademark of Byk-Chemie GmbH

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiangli Wang et al.
ChemSusChem, 5(7), 1307-1312 (2012-04-03)
Y(2)O(3):Er(3+) nanorods are synthesized by means of a hydrothermal method and then introduced into a TiO(2) electrode in a dye-sensitized solar cell (DSSC). Y(2)O(3):Er(3+) improves infrared light harvest via up-conversion luminescence and increases the photocurrent of the DSSC. The rare
Cheol Jang et al.
Optics express, 20(3), 2143-2148 (2012-02-15)
We demonstrate the optical characteristics of YVO4:Eu3+ phosphor in close proximity to Ag nanofilm to create a highly efficient emitting layer in mirror-type self-emissive displays. The propagating surface plasmon mode induced between the dielectric layer (MgO) and the Ag nanofilm
Masoud Allahkarami et al.
Dental materials : official publication of the Academy of Dental Materials, 27(12), 1279-1284 (2011-10-14)
Chipping failures observed clinically in bilayer systems of porcelain and zirconia restorations should be coupled with a monoclinic to tetragonal phase transformation in the zirconia layer due to the high compressive stress. Phase transformations were mapped using 2D micro X-ray
Eun-Ok Oh et al.
Advanced materials (Deerfield Beach, Fla.), 24(25), 3373-3377 (2012-06-01)
An extremely thin bilayer electrolyte consisting of yttria-stabilized zirconia (YSZ) and gadolinia-doped ceria (GDC) is successfully fabricated on a sintered NiO-YSZ substrate. Major processing flaws are effectively eliminated by applying local constraints to YSZ nanoparticles, and excellent open circuit voltage
Bipin Kumar Gupta et al.
Small (Weinheim an der Bergstrasse, Germany), 8(19), 3028-3034 (2012-07-19)
Highly luminescent-paramagnetic nanophosphors have a seminal role in biotechnology and biomedical research due to their potential applications in biolabeling, bioimaging, and drug delivery. Herein, the synthesis of high-quality, ultrafine, europium-doped yttrium oxide nanophosphors (Y(1.9)O(3):Eu(0.1)(3+)) using a modified sol-gel technique is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service