Select a Size
Select a Size
About This Item
Assay
96%
refractive index
n20/D 1.4910 (lit.)
bp
45-52 °C/0.5 mmHg (lit.)
density
1.020 g/mL at 25 °C (lit.)
SMILES string
COC1=CCC=C(C1)OC
InChI
1S/C8H12O2/c1-9-7-4-3-5-8(6-7)10-2/h4-5H,3,6H2,1-2H3
InChI key
MDBSJHGNFNKPTA-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
185.0 °F - closed cup
Flash Point(C)
85 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service