Skip to Content
Merck
All Photos(1)

Key Documents

639109

Sigma-Aldrich

2-Amino-3-pyridinecarboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

98-102 °C (lit.)

SMILES string

[H]C(=O)c1cccnc1N

InChI

1S/C6H6N2O/c7-6-5(4-9)2-1-3-8-6/h1-4H,(H2,7,8)

InChI key

NXMFJCRMSDRXLD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanhong Liu et al.
Dalton transactions (Cambridge, England : 2003), 48(36), 13834-13840 (2019-09-05)
A novel aldehyde- and amino-functionalized luminescent metal-organic framework, Cd-TCHO, was constructed through the solvothermal reaction of 4,4',4''-tricarboxytriphenylamine, 2-amino-3-pyridinecarboxaldehyde and cadmium nitrate and was characterized. Post-synthetically oxidizing the aldehyde groups into carboxylate groups afforded a new complex, Cd-TCOOH, and this successful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service