Skip to Content
Merck
All Photos(1)

Documents

614017

Sigma-Aldrich

Vanillin-5-d1

≥90 atom % D, ≥99% (CP)

Synonym(s):

4-Hydroxy-3-methoxybenzaldehyde-5-d1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8DH7O3
Molecular Weight:
153.15
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

isotopic purity

≥90 atom % D

Assay

≥99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

mass shift

M+1

SMILES string

[2H]c1cc(C=O)cc(OC)c1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3/i3D

InChI key

MWOOGOJBHIARFG-WFVSFCRTSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service