Skip to Content
Merck
All Photos(1)

Documents

606693

Sigma-Aldrich

Tetrachloroethylene-13C2

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl213C=13CCl2
CAS Number:
Molecular Weight:
167.82
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

121 °C (lit.)

mp

-22 °C (lit.)

density

1.630 g/mL at 25 °C

mass shift

M+2

SMILES string

Cl\[13C](Cl)=[13C](/Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6/i1+1,2+1

InChI key

CYTYCFOTNPOANT-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rocio Aranda-Rodriguez et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 992, 76-85 (2015-05-13)
Humans are continuously exposed to volatile organic compounds (VOCs) as these chemicals are ubiquitously present in most indoor and outdoor environments. In order to assess recent exposure to VOCs for population-based studies, VOCs are measured in the blood of participants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service