Skip to Content
Merck
All Photos(1)

Key Documents

590126

Sigma-Aldrich

Benzyl 2-propylacrylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH2CH2CH3)CO2CH2C6H5
CAS Number:
Molecular Weight:
204.26
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

storage temp.

2-8°C

SMILES string

[H]\C([H])=C(\CCC)C(=O)OCc1ccccc1

InChI

1S/C13H16O2/c1-3-7-11(2)13(14)15-10-12-8-5-4-6-9-12/h4-6,8-9H,2-3,7,10H2,1H3

InChI key

VVMMMVHSJPQPMA-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service