Skip to Content
Merck
All Photos(1)

Key Documents

588180

Sigma-Aldrich

1-Dimethylamino-2-propylamine

98%

Synonym(s):

N,N-Dimethylpropylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2N(CH3)2
CAS Number:
Molecular Weight:
102.18
Beilstein:
969185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.421 (lit.)

bp

113 °C (lit.)

density

0.792 g/mL at 25 °C (lit.)

SMILES string

CC(N)CN(C)C

InChI

1S/C5H14N2/c1-5(6)4-7(2)3/h5H,4,6H2,1-3H3

InChI key

RRQHLOZQFPWDCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service